6,7-Dihydro-5h-cyclopenta[b]pyridin-7-yl acetate 95%

Lieferant: Apollo Scientific

OR919283-1G
APOSOR919283-1GEA 104 EUR
APOSOR919283-1G
6,7-Dihydro-5h-cyclopenta[b]pyridin-7-yl acetate 95%
6,7-Dihydro-5h-cyclopenta[b]pyridin-7-yl acetate
Formel: C₁₀H₁₁NO₂
Molecular Weight: 177,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19704614
CAS-Nummer: 90685-59-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR