3-Acetyl-1-methylpyrrol 95%

Lieferant: Apollo Scientific

Synonyme: 3-Acetyl-1-methylpyrrole

OR919291-5G
APOSOR919291-5GEA 154 EUR
APOSOR919291-5G
3-Acetyl-1-methylpyrrol 95%
3-Acetyl-1-methylpyrrol
Formel: C₇H₉NO
Molecular Weight: 123,15 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 932-62-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR