Methyl 3-amino-2,4-dimethylbenzoate 95%

Lieferant: Apollo Scientific

OR919396-1G
APOSOR919396-1GEA 116 EUR
APOSOR919396-1G
Methyl 3-amino-2,4-dimethylbenzoate 95%
Methyl 3-amino-2,4-dimethylbenzoate
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179,216 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22494937
CAS-Nummer: 24812-89-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR