2,5-Dichlorterephthalonitril 97%

Lieferant: Apollo Scientific

Synonyme: 1,4-Dichloro-2,5-dicyanobenzene

OR919416-5G
APOSOR919416-5GEA 200 EUR
APOSOR919416-5G
2,5-Dichlorterephthalonitril 97%
2,5-Dichloroterephthalonitrile
Formel: C₈H₂Cl₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00059584
CAS-Nummer: 1897-43-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR