4-(Dimethylamino)-1-butanol 98%

Lieferant: Apollo Scientific

OR919427-100G OR919427-25G
APOSOR919427-100GEA 0 EUR
APOSOR919427-100G APOSOR919427-25G
4-(Dimethylamino)-1-butanol 98%
4-(Dimethylamino)-1-butanol
Formel: C₆H₁₅NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00020624
CAS-Nummer: 13330-96-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR