4-Chloro-6,7-dihydro-5h-cyclopenta[d]pyrimidin-2-amine 95%
Lieferant: Apollo Scientific
OR919512-1G
APOSOR919512-1GEA
864
EUR
APOSOR919512-1G
4-Chloro-6,7-dihydro-5h-cyclopenta[d]pyrimidin-2-amine 95%
4-Chloro-6,7-dihydro-5h-cyclopenta[d]pyrimidin-2-amine
Formel:
C₇H₈ClN₃ Molecular Weight: 169,612 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18916997 CAS-Nummer: 5461-89-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...