2,3-Dimethoxybenzylalkohol 95%

Lieferant: Apollo Scientific

Synonyme: (2,3-Dimethoxyphenyl)methanol

OR919570-25G
APOSOR919570-25GEA 114 EUR
APOSOR919570-25G
2,3-Dimethoxybenzylalkohol 95%
2,3-Dimethoxybenzylalkohol
Formel: C₉H₁₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00004612
CAS-Nummer: 5653-67-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR