2,4,4-Trimethyl-2-oxazolin 98%

Lieferant: Apollo Scientific

Synonyme: 2,4,4-Trimethyl-4,5-dihydrooxazole

OR919620-25G
APOSOR919620-25GEA 74.1 EUR
APOSOR919620-25G
2,4,4-Trimethyl-2-oxazolin 98%
2,4,4-Trimethyl-2-oxazolin
Formel: C₆H₁₁NO
Molecular Weight: 113.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00005301
CAS-Nummer: 1772-43-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR