6-Bromo-3-hydroxypyrazine-2-carboxamide

Lieferant: Apollo Scientific

OR919629-5G
APOSOR919629-5GEA 66.7 EUR
APOSOR919629-5G
6-Bromo-3-hydroxypyrazine-2-carboxamide
6-Bromo-3-hydroxypyrazine-2-carboxamide
Formel: C₅H₄BrN₃O₂
Molecular Weight: 218.008 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD20528005
CAS-Nummer: 259793-88-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR