N-(4-Hydroxyphenyl)Methacrylamid 98%
Lieferant: Apollo Scientific
OR919641-25G
OR919641-5G
APOSOR919641-25GEA
287
EUR
APOSOR919641-25G
APOSOR919641-5G
N-(4-Hydroxyphenyl)Methacrylamid 98%
N-(4-Hydroxyphenyl)Methacrylamid
Formel:
C₁₀H₁₁NO₂ Molecular Weight: 177.20 g/mol Lagertemperatur: Raumtemperatur |
CAS-Nummer:
19243-95-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...