(R)-tert-Butyl 2-ethylpyrrolidine-1-carboxylate 95%

Lieferant: Apollo Scientific

OR919715-1G
APOSOR919715-1GEA 101 EUR
APOSOR919715-1G
(R)-tert-Butyl 2-ethylpyrrolidine-1-carboxylate 95%
(R)-tert-Butyl 2-ethylpyrrolidine-1-carboxylate
Formel: C₁₁H₂₁NO₂
Molecular Weight: 199,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24466674
CAS-Nummer: 876617-06-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR