2-(4-Methylphenoxy)ethyl-acetat 95%

Lieferant: Apollo Scientific

Synonyme: 2-(p-Tolyloxy)ethyl acetate, Ethylene Glycol Mono-p-tolyl Ether Acetate, Acetic Acid 2-p-Tolyloxyethyl Ester, 2-p-Tolyloxyethyl Acetate

OR919731-10G OR919731-1G OR919731-5G
APOSOR919731-10GEA 134 EUR
APOSOR919731-10G APOSOR919731-1G APOSOR919731-5G
2-(4-Methylphenoxy)ethyl-acetat 95%
2-(4-Methylphenoxy)ethyl-acetat
Formel: C₁₁H₁₄O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00059337
CAS-Nummer: 6807-11-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR