S-Furfurylthioformiat 95%

Lieferant: Apollo Scientific

OR919771-5G
APOSOR919771-5GEA 473 EUR
APOSOR919771-5G
S-Furfurylthioformiat 95%
S-Furfurylthioformiat
Formel: C₆H₆O₂S
Lagertemperatur: Kühlschrank
MDL: MFCD00209507
CAS-Nummer: 59020-90-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR