Ethyl 3-cyclopropylpyrrole-2-carboxylate 95%
Lieferant: Apollo Scientific
Synonyme:
Ethyl 3-cyclopropyl-1H-pyrrole-2-carboxylate
OR919811-1G
APOSOR919811-1GEA
237
EUR
APOSOR919811-1G
Ethyl 3-cyclopropylpyrrole-2-carboxylate 95%
Ethyl 3-cyclopropylpyrrole-2-carboxylate
Formel:
C₁₀H₁₃NO₂ Molecular Weight: 179.216 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12924310 CAS-Nummer: 1123725-69-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...