2,3,4-Trimethyl-3-pentanol 95%
Lieferant: Apollo Scientific
Synonyme:
2,3,4-Trimethylpentan-3-ol
OR919878-5G
APOSOR919878-5GEA
200
EUR
APOSOR919878-5G
2,3,4-Trimethyl-3-pentanol 95%
2,3,4-Trimethyl-3-pentanol
Formel:
C₈H₁₈O Molecular Weight: 130.23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00048319 CAS-Nummer: 3054-92-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...