3-Phenylpropionitril 95%
Lieferant: Apollo Scientific
Synonyme:
Hydrocinnamonitrile
OR919888-25G
APOSOR919888-25GEA
105
EUR
APOSOR919888-25G
3-Phenylpropionitril 95%
3-Phenylpropionitril
Formel:
C₉H₉N Molecular Weight: 131,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001961 CAS-Nummer: 645-59-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...