Methyl 2-(chloromethyl)acrylate 95%

Lieferant: Apollo Scientific

OR919913-1G OR919913-5G
APOSOR919913-1GEA 166 EUR
APOSOR919913-1G APOSOR919913-5G
Methyl 2-(chloromethyl)acrylate 95%
Methyl 2-(chloromethyl)acrylate
Formel: C₅H₇ClO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11617809
CAS-Nummer: 922-15-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR