3,5-Dimethyl-3-hexanol 95%

Lieferant: Apollo Scientific

OR919924-5G
APOSOR919924-5GEA 226 EUR
APOSOR919924-5G
3,5-Dimethyl-3-hexanol 95%
3,5-Dimethyl-3-hexanol
Formel: C₈H₁₈O
Molecular Weight: 130,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00021823
CAS-Nummer: 4209-91-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR