5-(4-Ethynylphenyl)-1,3-oxazole 97%

Lieferant: Apollo Scientific
Achtung

OR9200-1G OR9200-250MG
APOSOR9200-1GEA 452 EUR
APOSOR9200-1G APOSOR9200-250MG
5-(4-Ethynylphenyl)-1,3-oxazole 97%
5-(4-Ethynylphenyl)-1,3-oxazole
Formel: C₁₁H₇NO
Lagertemperatur: Raumtemperatur
MDL: MFCD08435847
CAS-Nummer: 501944-63-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR