4-sec-Butylcyclohexanamin (cis/trans-Gemisch) 95%

Lieferant: Apollo Scientific

OR920000-5G
APOSOR920000-5GEA 560 EUR
APOSOR920000-5G
4-sec-Butylcyclohexanamin (cis/trans-Gemisch) 95%
4-sec-Butylcyclohexylamine (cis- and trans- mixture)
Formel: C₁₀H₂₁N
Lagertemperatur: Raumtemperatur
CAS-Nummer: 25834-93-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR