N-(2-Acetyl-phenyl)-acetamide 98%

Lieferant: Apollo Scientific

OR920016-1G OR920016-5G
APOSOR920016-1GEA 0 EUR
APOSOR920016-1G APOSOR920016-5G
N-(2-Acetyl-phenyl)-acetamide 98%
N-(2-Acetyl-phenyl)-acetamide
Formel: C₁₀H₁₁NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00032277
CAS-Nummer: 5234-26-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR