1,4-Benzoquinone dioxime 95%

Lieferant: Apollo Scientific

OR920023-100G OR920023-250G OR920023-25G
APOSOR920023-100GEA 226 EUR
APOSOR920023-100G APOSOR920023-250G APOSOR920023-25G
1,4-Benzoquinone dioxime 95%
1,4-Benzoquinone dioxime
Formel: C₆H₆N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00063636
CAS-Nummer: 105-11-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR