Methylhydrogenglutarat 95%

Lieferant: Apollo Scientific

Synonyme: 5-Methoxy-5-oxopentanoic acid

OR920036-25G
APOSOR920036-25GEA 60.9 EUR
APOSOR920036-25G
Methylhydrogenglutarat 95%
Methylhydrogenglutarat
Formel: C₆H₁₀O₄
Molecular Weight: 146.1412 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00004409
CAS-Nummer: 1501-27-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR