(1S,2R)-2-(Dibutylamino)-1-phenyl-1-propanol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (-)-N,N-Dibutylnorephedrin, (-)-alpha-[1-(Dibutylamino)ethyl]benzyl Alcohol

OR920038-5G
APOSOR920038-5GEA 277 EUR
APOSOR920038-5G
(1S,2R)-2-(Dibutylamino)-1-phenyl-1-propanol 98%
(1S,2R)-2-(Dibutylamino)-1-phenyl-1-propanol
Formel: C₁₇H₂₉NO
Molecular Weight: 263,43 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00142695
CAS-Nummer: 114389-70-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR