(±)-1-Heptin-3-ol 97%
Lieferant: Apollo Scientific
Synonyme:
Hept-1-yn-3-ol
OR920099-5G
APOSOR920099-5GEA
55.1
EUR
APOSOR920099-5G
(±)-1-Heptin-3-ol 97%
(±)-1-Heptin-3-ol
Formel:
C₇H₁₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041597 CAS-Nummer: 7383-19-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...