Spiro[3.3]heptan-2-amine hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR920126-100MG
APOSOR920126-100MGEA 63.8 EUR
APOSOR920126-100MG
Spiro[3.3]heptan-2-amine hydrochloride 97%
Spiro[3.3]heptan-2-amine hydrochloride
Formel: C₇H₁₃N·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD22689925
CAS-Nummer: 1416439-08-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR