4-Butylcyclohexanol (cis/trans-Gemisch) 95%

Lieferant: Apollo Scientific

OR920133-5G
APOSOR920133-5GEA 142 EUR
APOSOR920133-5G
4-Butylcyclohexanol (cis/trans-Gemisch) 95%
4-Butylcyclohexanol (cis- and trans- mixture)
Formel: C₁₀H₂₀O
Lagertemperatur: Raumtemperatur
MDL: MFCD01321147
CAS-Nummer: 70568-60-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR