D-Allo-isoleucine ethyl ester hydrochloride 95%

Lieferant: Apollo Scientific

Synonyme: D-allo-Isoleucine Ethyl Ester Hydrochloride, H-D-Allo-Ile-OEt·HCl

OR920149-1G
APOSOR920149-1GEA 168 EUR
APOSOR920149-1G
D-Allo-isoleucine ethyl ester hydrochloride 95%
D-Allo-isoleucine ethyl ester hydrochloride
Formel: C₈H₁₇NO₂·ClH
Molecular Weight: 195,687 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26523292
CAS-Nummer: 315700-65-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR