8-Hydroxy-7-propylquinoline 98%

Lieferant: Apollo Scientific

Synonyme: 7-Propyloxine, 7-Propyl-8-quinolinol

OR920199-1G
APOSOR920199-1GEA 522 EUR
APOSOR920199-1G
8-Hydroxy-7-propylquinoline 98%
8-Hydroxy-7-propylquinoline
Formel: C₁₂H₁₃NO
Molecular Weight: 187,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00059755
CAS-Nummer: 58327-60-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR