4-(4-Isocyanatopyridin-2-yl)morpholine 90%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Isocyanato-2-(morpholin-4-yl)pyridine, 2-(Morpholin-4-yl)pyridin-4-yl isocyanate

OR9203-250MG
APOSOR9203-250MGEA 118 EUR
APOSOR9203-250MG
4-(4-Isocyanatopyridin-2-yl)morpholine 90%
4-(4-Isocyanatopyridin-2-yl)morpholine

When melted, may contain small amount of sediment

Formel: C₁₀H₁₁N₃O₂
Molecular Weight: 205,213 g/mol
Schmelzpunkt: 67…69 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08271914
CAS-Nummer: 876316-43-7
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR