1-Ethyl-3-pyrrolidinol 95%

Lieferant: Apollo Scientific

Synonyme: 1-Ethyl-3-hydroxypyrrolidine

OR920330-5G
APOSOR920330-5GEA 56.7 EUR
APOSOR920330-5G
1-Ethyl-3-pyrrolidinol 95%
1-Ethyl-3-pyrrolidinol
Formel: C₆H₁₃NO
Molecular Weight: 115.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00003179
CAS-Nummer: 30727-14-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR