4-Chloro-1-methyl-1H-imidazo[4,5-c]pyridine 95%

Lieferant: Apollo Scientific

OR920336-1G
APOSOR920336-1GEA 121 EUR
APOSOR920336-1G
4-Chloro-1-methyl-1H-imidazo[4,5-c]pyridine 95%
4-Chloro-1-methyl-1H-imidazo[4,5-c]pyridine
Formel: C₇H₆ClN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00222101
CAS-Nummer: 50432-68-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR