Trans-1,2-cyclopropane-dicarboxylic acid mono ethyl ester 95%
Lieferant: Apollo Scientific
OR920382-1G
APOSOR920382-1GEA
632
EUR
APOSOR920382-1G
Trans-1,2-cyclopropane-dicarboxylic acid mono ethyl ester 95%
Trans-1,2-cyclopropane-dicarboxylic acid mono ethyl ester
Formel:
C₇H₁₀O₄ Molecular Weight: 158,152 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD26383405 CAS-Nummer: 31420-66-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...