(4-Methoxyphenyl)essigsäureethylester 95%

Lieferant: Apollo Scientific

Synonyme: Ethyl-(4-methoxyphenyl)acetat

OR920393-100G
APOSOR920393-100GEA 133 EUR
APOSOR920393-100G
(4-Methoxyphenyl)essigsäureethylester 95%
(4-Methoxyphenyl)essigsäureethylester
Formel: C₁₁H₁₄O₃
Molecular Weight: 194.23 g/mol
Siedepunkt: 138
Lagertemperatur: Raumtemperatur
MDL: MFCD00040760
CAS-Nummer: 14062-18-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR