2,4-Dichloro-3-hydroxybenzaldehyde 95%
Lieferant: Apollo Scientific
OR920426-1G
APOSOR920426-1GEA
288
EUR
APOSOR920426-1G
2,4-Dichloro-3-hydroxybenzaldehyde 95%
2,4-Dichloro-3-hydroxybenzaldehyde
Formel:
C₇H₄Cl₂O₂ Molecular Weight: 191,011 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22199563 CAS-Nummer: 56962-13-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...