1-Methylazepan-4-ol 95%

Lieferant: Apollo Scientific

Synonyme: 1-Methyl-4-azepanol hydrochloride

OR920530-1G
APOSOR920530-1GEA 144 EUR
APOSOR920530-1G
1-Methylazepan-4-ol 95%
1-Methylazepan-4-ol
Formel: C₇H₁₅NO
Molecular Weight: 129,2000 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00190597
CAS-Nummer: 19065-49-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR