2-Methylimidazo[1,2-b]pyridazin-6-amine 95%

Lieferant: Apollo Scientific

OR920652-1G
APOSOR920652-1GEA 220 EUR
APOSOR920652-1G
2-Methylimidazo[1,2-b]pyridazin-6-amine 95%
2-Methylimidazo[1,2-b]pyridazin-6-amine
Formel: C₇H₈N₄
Lagertemperatur: Raumtemperatur
MDL: MFCD19211982
CAS-Nummer: 154704-35-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR