2-(Fur-2-yl)benzeneboronic acid, pinacol ester 97%
Lieferant: Apollo Scientific
Synonyme:
2-[2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]furan, 2-[2-(Fur-2-yl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
OR9207-250MG
APOSOR9207-250MGEA
277
EUR
APOSOR9207-250MG
2-(Fur-2-yl)benzeneboronic acid, pinacol ester 97%
2-(Fur-2-yl)benzeneboronic acid, pinacol ester
Formel:
C₁₆H₁₉BO₃ Molecular Weight: 270,131 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08435879 CAS-Nummer: 876316-28-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...