1-Acetyl-1H-1,2,3-triazolo[4,5-b]pyridine 95%

Lieferant: Apollo Scientific

OR920701-1G OR920701-250MG OR920701-5G
APOSOR920701-1GEA 191 EUR
APOSOR920701-1G APOSOR920701-250MG APOSOR920701-5G
1-Acetyl-1H-1,2,3-triazolo[4,5-b]pyridine 95%
1-Acetyl-1H-1,2,3-triazolo[4,5-b]pyridine
Formel: C₇H₆N₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD00075383
CAS-Nummer: 107866-54-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR