S-Phenylthioacetat 95%

Lieferant: Apollo Scientific

OR920718-25G
APOSOR920718-25GEA 89.9 EUR
APOSOR920718-25G
S-Phenylthioacetat 95%
S-Phenylthioacetat
Formel: C₈H₈OS
Molecular Weight: 152,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00008752
CAS-Nummer: 934-87-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR