4,6-Dichloropyridine-2-carbonitrile 98%

Lieferant: Apollo Scientific

Synonyme: 4,6-Dichloropicolinonitrile

OR920721-1G
APOSOR920721-1GEA 66.7 EUR
APOSOR920721-1G
4,6-Dichloropyridine-2-carbonitrile 98%
4,6-Dichloropyridine-2-carbonitrile
Formel: C₆H₂Cl₂N₂
Molecular Weight: 172,999 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22684690
CAS-Nummer: 40314-71-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR