3-Ethoxy-N,N-diethylanilin 95%

Lieferant: Apollo Scientific

Synonyme: N,N-Diethyl-m-phenetidine

OR920744-25G
APOSOR920744-25GEA 105 EUR
APOSOR920744-25G
3-Ethoxy-N,N-diethylanilin 95%
3-Ethoxy-N,N-diethylanilin
Formel: C₁₂H₁₉NO
Molecular Weight: 193.29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00043936
CAS-Nummer: 1864-92-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR