(S)-(+)-1-(2-Pyrrolidinylmethyl)pyrrolidine 95%

Lieferant: Apollo Scientific

OR920747-1G
APOSOR920747-1GEA 82.8 EUR
APOSOR920747-1G
(S)-(+)-1-(2-Pyrrolidinylmethyl)pyrrolidine 95%
(S)-(+)-1-(2-Pyrrolidinylmethyl)pyrrolidine
Formel: C₉H₁₈N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00013441
CAS-Nummer: 51207-66-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR