Diisobutylamine hydrochloride 95%

Lieferant: Apollo Scientific

OR920752-25G
APOSOR920752-25GEA 0 EUR
APOSOR920752-25G
Diisobutylamine hydrochloride 95%
Diisobutylamine hydrochloride
Formel: C₈H₁₉N
Lagertemperatur: Raumtemperatur
MDL: MFCD00060151
CAS-Nummer: 18251-82-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR