4-Oxa-7-azaspiro[2.5]octane hydrochloride

Lieferant: Apollo Scientific

OR920787-1G
APOSOR920787-1GEA 426 EUR
APOSOR920787-1G
4-Oxa-7-azaspiro[2.5]octane hydrochloride
4-Oxa-7-azaspiro[2.5]octane hydrochloride
Formel: C₆H₁₁NO·ClH
Molecular Weight: 149.619 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21606609
CAS-Nummer: 1427195-23-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR