1-Butyl-4-nitrobenzol 95%
Lieferant: Apollo Scientific
OR920802-5G
APOSOR920802-5GEA
451
EUR
APOSOR920802-5G
1-Butyl-4-nitrobenzol 95%
1-Butyl-4-nitrobenzene
Formel:
C₁₀H₁₃NO₂ Lagertemperatur: Raumtemperatur |
CAS-Nummer:
20651-75-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...