9-Oxabicyclo[6.1.0]-4-nonen 98%

Lieferant: Apollo Scientific

OR920820-25G OR920820-5G
APOSOR920820-25GEA 666 EUR
APOSOR920820-25G APOSOR920820-5G
9-Oxabicyclo[6.1.0]-4-nonen 98%
9-Oxabicyclo[6.1.0]-4-nonen
Formel: C₈H₁₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00086420
CAS-Nummer: 637-90-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR