4-Amino-1-methyl-1H-pyrazole-3-carbonitrile 98%

Lieferant: Apollo Scientific

Synonyme: 4-Amino-1-methyl-1h-pyrazole-3-carbonitrile

OR920854-1G
APOSOR920854-1GEA 281 EUR
APOSOR920854-1G
4-Amino-1-methyl-1H-pyrazole-3-carbonitrile 98%
4-Amino-1-methyl-1H-pyrazole-3-carbonitrile
Formel: C₅H₆N₄
Molecular Weight: 122,128 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19215684
CAS-Nummer: 1201935-84-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR