4-Hydroxy-1-cyclohexanecarboxylic acid-δ-lactone 98%
Lieferant: Apollo Scientific
Synonyme:
3,6-Ethanotetrahydropyran-2-one, 2-Oxabicyclo[2.2.2]octan-3-one
OR920857-5G
APOSOR920857-5GEA
328
EUR
APOSOR920857-5G
4-Hydroxy-1-cyclohexanecarboxylic acid-δ-lactone 98%
4-Hydroxy-1-cyclohexanecarboxylic acid-delta-lactone
Formel:
C₇H₁₀O₂ Molecular Weight: 126,16 g/mol Siedepunkt: 123 °C/11mmHg Schmelzpunkt: 128 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06797111 CAS-Nummer: 4350-84-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...