1-(4-Hydroxy-phenyl)-cyclopropanecarboxylic acid 95%
Lieferant: Apollo Scientific
OR920881-1G
APOSOR920881-1GEA
478
EUR
APOSOR920881-1G
1-(4-Hydroxy-phenyl)-cyclopropanecarboxylic acid 95%
1-(4-Hydroxy-phenyl)-cyclopropanecarboxylic acid
Formel:
C₁₀H₁₀O₃ Molecular Weight: 178,185 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19441896 CAS-Nummer: 869970-25-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...